ENA447958297 Enamine

5-methyl-1-[(pyridin-4-yl)methyl]-1H-1,2,3-triazole-4-carboxylic acid hydrochloride

  • MDL number MFCD28397701
  • Cas Number : 1803601-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210134
Availability details for
Quantity
  • Availability
  • Add to Cart