ENA447958299 Enamine

1-[2-chloro-4-(trifluoromethyl)phenyl]piperazine hydrochloride

  • MDL number MFCD28397703
  • Cas Number : 1803571-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210137
Availability details for
Quantity
  • Availability
  • Add to Cart