ENA447958301 Enamine

1-(2-phenyl-1,3-thiazol-4-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28397706
  • Cas Number : 1803606-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210206
Availability details for
Quantity
  • Availability
  • Add to Cart