ENA447958302 Enamine

octahydropyrrolo[1,2-a]piperazine-1-carboxylic acid dihydrochloride

  • MDL number MFCD28397708
  • Cas Number : 1803566-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210210
Availability details for
Quantity
  • Availability
  • Add to Cart