ENA447958307 Enamine

2-[(azetidin-3-yl)formamido]-2-ethylbutanamide hydrochloride

  • MDL number MFCD28397715
  • Cas Number : 1803588-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210272
Availability details for
Quantity
  • Availability
  • Add to Cart