ENA447958310 Enamine

(1-fluorocycloheptyl)methanesulfonyl chloride

  • MDL number MFCD28397719
  • Cas Number : 1803565-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210294
Availability details for
Quantity
  • Availability
  • Add to Cart