ENA447958312 Enamine

(pyrrolidin-2-yl)methyl N-cyclohexylcarbamate hydrochloride

  • MDL number MFCD28397722
  • Cas Number : 1803604-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210718
Availability details for
Quantity
  • Availability
  • Add to Cart