ENA447958321 Enamine

(piperidin-2-yl)methyl N,N-diethylcarbamate hydrochloride

  • MDL number MFCD28397731
  • Cas Number : 1803572-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210802
Availability details for
Quantity
  • Availability
  • Add to Cart