ENA447958323 Enamine

(piperidin-2-yl)methyl 3-methylpiperidine-1-carboxylate hydrochloride

  • MDL number MFCD28397733
  • Cas Number : 1803588-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210806
Availability details for
Quantity
  • Availability
  • Add to Cart