ENA447958325 Enamine

(piperidin-2-yl)methyl pyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD28397735
  • Cas Number : 1803589-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210809
Availability details for
Quantity
  • Availability
  • Add to Cart