ENA447958328 Enamine

2,6-dichloro-N,N-diethyl-3,5-dimethoxybenzamide

  • MDL number MFCD28397738
  • Cas Number : 1803599-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210864
Availability details for
Quantity
  • Availability
  • Add to Cart