ENA447958331 Enamine

1-[(tert-butoxy)carbonyl]-4-methanesulfonylpiperidine-4-carboxylic acid

  • MDL number MFCD28397741
  • Cas Number : 1803588-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210869
Availability details for
Quantity
  • Availability
  • Add to Cart