ENA447958333 Enamine

propan-2-yl 4-(benzyloxy)-2,2-dimethylbutanoate

  • MDL number MFCD28397743
  • Cas Number : 1803606-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210876
Availability details for
Quantity
  • Availability
  • Add to Cart