ENA447958334 Enamine

(piperidin-2-yl)methyl azepane-1-carboxylate hydrochloride

  • MDL number MFCD28397744
  • Cas Number : 1803572-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210877
Availability details for
Quantity
  • Availability
  • Add to Cart