ENA447958335 Enamine

3-(methylamino)-4-phenylbutanoic acid hydrochloride

  • MDL number MFCD28397745
  • Cas Number : 60546-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210881
Availability details for
Quantity
  • Availability
  • Add to Cart