ENA447958337 Enamine

2-[(2-ethylpyrrolidin-2-yl)formamido]pent-4-enoic acid hydrochloride

  • MDL number MFCD28397747
  • Cas Number : 1803570-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210896
Availability details for
Quantity
  • Availability
  • Add to Cart