ENA447958338 Enamine

2-amino-4-(2-aminoethoxy)butanoic acid dihydrochloride

  • MDL number MFCD28397748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210898
Availability details for
Quantity
  • Availability
  • Add to Cart