ENA447958339 Enamine

3-(aminomethyl)pyrrolidine-1-carboxamide dihydrochloride

  • MDL number MFCD28397749
  • Cas Number : 1803586-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210933
Availability details for
Quantity
  • Availability
  • Add to Cart