ENA447958341 Enamine

4-(oxan-4-yl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD28397751
  • Cas Number : 1803601-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210965
Availability details for
Quantity
  • Availability
  • Add to Cart