ENA447958343 Enamine

potassium 3-(phenylcarbamoyl)benzoate

  • MDL number MFCD28397753
  • Cas Number : 1803589-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210977
Availability details for
Quantity
  • Availability
  • Add to Cart