ENA447958346 Enamine

5-(piperidin-2-yl)-2,3-dihydro-1H-1,2,4-triazol-3-one hydrochloride

  • MDL number MFCD28397757
  • Cas Number : 1803606-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211054
Availability details for
Quantity
  • Availability
  • Add to Cart