ENA447958349 Enamine

2,2-difluoro-2-[3-(propan-2-yl)-1H-pyrazol-5-yl]ethan-1-amine

  • MDL number MFCD28397761

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211096
Availability details for
Quantity
  • Availability
  • Add to Cart