ENA447958354 Enamine

[1-(dimethylamino)cyclopropyl]methanol hydrochloride

  • MDL number MFCD28397767
  • Cas Number : 1803606-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211234
Availability details for
Quantity
  • Availability
  • Add to Cart