ENA447958357 Enamine

5-methyl-1-(piperidin-4-yl)-1H-pyrazole-4-carboxylic acid dihydrochloride

  • MDL number MFCD28397771
  • Cas Number : 1803567-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211366
Availability details for
Quantity
  • Availability
  • Add to Cart