ENA447958362 Enamine

N-hydroxy-2-methylquinoline-4-carboxamide

  • MDL number MFCD09935718
  • Cas Number : 1016791-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208950
Availability details for
Quantity
  • Availability
  • Add to Cart