ENA447958368 Enamine

4-(2,3-dihydro-1H-indole-1-carbonyl)-1-[3-(methylsulfanyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD11723525
  • Cas Number : 1090029-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208417
Availability details for
Quantity
  • Availability
  • Add to Cart