ENA447958371 Enamine

[2-(propan-2-yl)-1,3-oxazol-4-yl]methanol

  • MDL number MFCD16556117
  • Cas Number : 162740-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208965
Availability details for
Quantity
  • Availability
  • Add to Cart