ENA447958373 Enamine

2-[(3-hydroxypropyl)amino]propanenitrile

  • MDL number MFCD10014403
  • Cas Number : 1018512-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208989
Availability details for
Quantity
  • Availability
  • Add to Cart