ENA447958381 Enamine

[2-(4-benzylpiperazin-1-yl)ethyl](methyl)amine

  • MDL number MFCD10697869
  • Cas Number : 157368-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208292
Availability details for
Quantity
  • Availability
  • Add to Cart