ENA447958385 Enamine

5-amino-2-fluoro-N-methylbenzene-1-sulfonamide

  • MDL number MFCD11621167
  • Cas Number : 1094887-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208426
Availability details for
Quantity
  • Availability
  • Add to Cart