ENA447958386 Enamine

1-(1-chloro-3-methylbutyl)-4-ethylbenzene

  • MDL number MFCD18353523
  • Cas Number : 1406246-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209058
Availability details for
Quantity
  • Availability
  • Add to Cart