ENA447958387 Enamine

2-(cyanosulfanyl)-1-(3,4-dihydroxyphenyl)ethan-1-one

  • MDL number MFCD22415334
  • Cas Number : 101714-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209059
Availability details for
Quantity
  • Availability
  • Add to Cart