ENA447958390 Enamine

methyl 2-(methoxymethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD23936699
  • Cas Number : 1512499-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209063
Availability details for
Quantity
  • Availability
  • Add to Cart