ENA447958394 Enamine

1-bromo-4-(2-chloroethyl)-2-fluorobenzene

  • MDL number MFCD24108151
  • Cas Number : 1370600-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209070
Availability details for
Quantity
  • Availability
  • Add to Cart