ENA447958395 Enamine

1-tert-butyl-4-(1-chloroethyl)benzene

  • MDL number MFCD11186417
  • Cas Number : 13372-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209072
Availability details for
Quantity
  • Availability
  • Add to Cart