ENA447958400 Enamine

1-(bromomethyl)-3-ethoxycyclobutane

  • MDL number MFCD28397621
  • Cas Number : 1694151-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209099
Availability details for
Quantity
  • Availability
  • Add to Cart