ENA447958408 Enamine

2-(aminomethyl)-4-bromophenol hydrochloride

  • MDL number MFCD11045214
  • Cas Number : 1803612-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208439
Availability details for
Quantity
  • Availability
  • Add to Cart