ENA447958410 Enamine

1-(4-bromo-2-methylphenyl)piperazine-2,5-dione

  • MDL number MFCD21115233
  • Cas Number : 1486833-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208440
Availability details for
Quantity
  • Availability
  • Add to Cart