ENA447958412 Enamine

methyl 2-amino-5-bromo-3,6-difluorobenzoate

  • MDL number MFCD24230476
  • Cas Number : 1549848-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208444
Availability details for
Quantity
  • Availability
  • Add to Cart