ENA447958414 Enamine

N-(2-methylpropyl)isoquinoline-5-sulfonamide

  • MDL number MFCD11986509
  • Cas Number : 84468-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209120
Availability details for
Quantity
  • Availability
  • Add to Cart