ENA447958416 Enamine

3-hydrazinyl-6-[3-(trifluoromethyl)phenyl]pyridazine

  • MDL number MFCD15522371
  • Cas Number : 76970-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208447
Availability details for
Quantity
  • Availability
  • Add to Cart