ENA447958442 Enamine

2,2,6,6-tetramethyl-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD01691457
  • Cas Number : 1005-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209174
Availability details for
Quantity
  • Availability
  • Add to Cart