ENA447958446 Enamine

2-{benzyl[(2,5-dimethylphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD01307798
  • Cas Number : 416893-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208576
Availability details for
Quantity
  • Availability
  • Add to Cart