ENA447958457 Enamine

3-tert-butyl-1-methylimidazolidine-2,4-dione

  • MDL number MFCD03923803
  • Cas Number : 905484-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209191
Availability details for
Quantity
  • Availability
  • Add to Cart