ENA447958463 Enamine

tert-butyl N-(trimethylsilyl)carbamate

  • MDL number MFCD24466841
  • Cas Number : 238096-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209203
Availability details for
Quantity
  • Availability
  • Add to Cart