ENA447958472 Enamine

5-cyclopropyl-1H-1,2,4-triazole-3-carboxamide

  • MDL number MFCD23886054
  • Cas Number : 1502673-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209214
Availability details for
Quantity
  • Availability
  • Add to Cart