ENA447958474 Enamine

N-(4-amino-2-chlorophenyl)-5-chloro-2-hydroxybenzamide

  • MDL number MFCD00569542
  • Cas Number : 10558-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-197
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209216
Availability details for
Quantity
  • Availability
  • Add to Cart