ENA447958476 Enamine

3-{methyl[2-(piperazin-1-yl)ethyl]amino}propanoic acid

  • MDL number MFCD18305336
  • Cas Number : 1405741-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209218
Availability details for
Quantity
  • Availability
  • Add to Cart