ENA447958481 Enamine

3-(phenylsulfanyl)pyridine

  • MDL number MFCD00234708
  • Cas Number : 28856-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209235
Availability details for
Quantity
  • Availability
  • Add to Cart