ENA447958483 Enamine

5-[(methylamino)methyl]-2,3-dihydro-1,3,4-oxadiazol-2-one hydrochloride

  • MDL number MFCD11109598
  • Cas Number : 1046079-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209251
Availability details for
Quantity
  • Availability
  • Add to Cart